Orders LinkContactUsLinkComponent
Test tubes, disposable
Test tubes, disposable
Supplier:   SEMADENI
Test tubes, disposable
Supplier:  SEMADENI
Certificates
Test tubes, disposable
Test tubes, disposable
2 Options Available Below
Product Details & Documents
PS, clear.
  • Good thermal resistance
  • Good chemical resistance
  • Robust, economical
Universal tubes for a wide range of applications.
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Availability
    Price
  • 10 ml
    100 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 17.90
    SFr. 17.90
    1 * 100 ST
       SFr. 17.90
  • 20 ml
    150 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 21.40
    SFr. 21.40
    1 * 100 ST
       SFr. 21.40