ContactUsLinkComponent
IP ActivFlo Biopsy II cassettes
IP ActivFlo Biopsy II cassettes
Catalog # 720-2651
Supplier:  LEICA HISTOLOGY
IP ActivFlo Biopsy II cassettes
Catalog # 720-2651
Supplier:  LEICA HISTOLOGY
Specifications
  • Colour:
    White
  • Environmentally Preferable:
  • Pk:
    1.000
Specifications
About this item
Surgipath ActivFlo Biopsy II cassettes feature a patented 'Fluidic Flow' pore design for maximum reagent exchange. With a 0,67 mm pore size, the cassettes include an innovative V-shape pore design that pulls fluid in and forces air out. The cassettes also feature sloped walls, creating a basket for small tissue management. These two piece cassettes with hinged lids have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
  • Available in taped packaging for easy printer loading