Orders LinkContactUsLinkComponent
Cooling Plates
Cooling Plates
Cooling plate, 480×280×33 mm (ext)
Catalog # 215-0248
Supplier:   SEMADENI
Cooling Plates
Catalog # 215-0248
Supplier:  SEMADENI
Cooling Plates
Cooling plate, 480×280×33 mm (ext)
Price
SFr. 110.00
Log in for availability
1 * 1 ST
Product Details & Documents
Cooling container, blue colour with white interior, suitable for transporting frozen or perishable products and laboratory products.
  • High quality foam of insulation
  • Hinged lid for tightly closing and two side handles
  • Stackable cooling plates
Use of cooling plate in the cover allows to effectively increase the cooling effect.
Specifications
  • Description:
    Cooling plate
  • Dimensions:
    480×280×33 mm (ext)
  • Seq:
    02