Order Entry
Switzerland
Orders LinkContactUsLinkComponent
LED ring light

LED ring light

VisiLED ring light, S 80-55 BF, WD: 45 - 95 mm, D: 66 mm
Catalog # ZEIS4170959255000
Supplier:   ZEISS
LED ring light
Catalog # ZEIS4170959255000
Supplier:  ZEISS
LED ring light
LED ring light

VisiLED ring light, S 80-55 BF, WD: 45 - 95 mm, D: 66 mm

Price
SFr. 1’176.10
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    VisiLED ring light, S 80-55 BF, WD: 45 - 95 mm, D: 66 mm
  • Seq:
    400