Clamps for standard ground joints
Nome del fornitore: MARIENFELD
Certificati
Informazioni su questo articolo
These clamps made of POM for standard ground joints are used to securely link the socket and the cone.
- Excellent chemical resistance
- Material avoids scratching on the glass surfaces
- Can be used at temperatures up to +150 °C
- Clips are colour coded for easy identification
Frequently Bought Together