Microcentrifuge tubes
NEOL7-4590
: NEOLAB
- Packed:1000 pcs/pk
- Type:with thread, without screw cap
- Version:non graduated, skirted, non sterile
- Capacity:0,5 ml
- Colour:Amber
- Pk:1.000
PP, amber coloured.
- Ungraduated
- Amber coloured to protect light sensitive substances
- Excellent chemical resistance
- Excellent temperature resistance
With thread.
: Screw caps must be ordered seperately.