Order Entry
Switzerland
ContactUsLinkComponent
Jars with knob lid
Jars with knob lid
Catalog # NEOL1-1183
Supplier:  NEOLAB
undefined
Jars with knob lid
Catalog # NEOL1-1183
Supplier:  NEOLAB

Specifications

  • Capacity:
    1200 ml
  • Height:
    120 mm
  • Neck-Ø:
    120 mm
  • Pk:
    1

Specifications

About this item

Jars, made of stainless steel, with knob lid.

  • Easy to clean
  • Resistant to chemicals
  • Excellent temperature resistance

Suitable for storing sensitive samples and pasty materials.