ContactUsLinkComponent
Lab markers, water resistant
Lab markers, water resistant
Catalog # NEOL2-5040
Supplier:  NEOLAB
undefined
Lab markers, water resistant
Catalog # NEOL2-5040
Supplier:  NEOLAB
Specifications
  • Colour:
    Black
  • Description:
    Laboratory marker, water resistant, fine tip
  • Pk:
    1
Specifications
About this item
Markers write on all materials used in a laboratory (as glass, plastics, porcelain, metal, wood, paper, etc.) and perform equally well on dry or moist, cold or warm surfaces.
  • Fine tip
  • Ink won't bleed when subjected to water or alcohol during normal use
  • Yet can be washed off