ContactUsLinkComponent
Cut resistant gloves, Showa 540D
Cut resistant gloves, Showa 540D
Supplier:  Showa
Certificates
undefined
Cut resistant gloves, Showa 540D
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-5858
  • 112-5857
  • 112-5856
  • 112-5860
  • 112-5859
  • Model:
  • Palm Fit 540D
  • Palm Fit 540D
  • Palm Fit 540D
  • Palm Fit 540D
  • Palm Fit 540D
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • HPPE
  • HPPE
  • HPPE
  • HPPE
  • HPPE
  • Finish:
  • Coating on palm
  • Coating on palm
  • Coating on palm
  • Coating on palm
  • Coating on palm
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4241B)
  • EN 388:2016 (4241B)
  • EN 388:2016 (4241B)
  • EN 388:2016 (4241B)
  • EN 388:2016 (4241B)