Orders LinkContactUsLinkComponent
Basic ESD Chairs
Basic ESD Chairs
 :   bimos
Basic ESD Chairs
 :  bimos
Basic ESD Chairs
Basic ESD Chairs
An ergonomic design, superb cleanroom properties and reliable ESD measures are a lot to ask from a workplace chair. Yet Cleanroom Basic ESD meets all these criteria and offers a high level of quality in the process.
  • Conductive high-comfort upholstery: choice of fabric or synthetic leather
  • Seat and backrest height adjustment
  • The frame is available in either black or light grey
  • Choice of three different mechanisms: contact backrest, contact backrest with seat tilt adjustment and synchronous mechanism with weight regulation
  • Steel components with conductive coating
  • Five volume conductive castors/glides
  • With five star base
Basic ESD has conductive covers. It features a sturdy five-legged base frame with flat lines, which is made from epoxyresin coated sectional steel tubing and provides good stability.
Certifications: Conform to DIN 68877, discharge and surface resistance according to EN 61340-5-1
German GS 'safety tested' certificate
Quality Management System DIN EN ISO 9001
Regulation EWG Nr. 1836/93 and
Environmental Management System DIN EN ISO 14001
Product Family Options
  • Description
    Height
    Foot style
    With
  • Basic ESD 2 chair, made from skai® synthetic leather, black, with contact backrest, backrest height: 430 mm
    470 - 610 mm
    Castors
    Five-legged base
    1 * 1 ST
    1 * 1 ST - SFr. 630.00
    SFr. 630.00
    1 * 1 ST
       SFr. 630.00
  • Basic ESD 2 chair, made from Duotec ESD fabric, black, with contact backrest with tilting seat, backrest height: 490 mm
    490 - 630 mm
    Castors
    Five-legged base
    1 * 1 ST
    1 * 1 ST - SFr. 776.00
    SFr. 776.00
    1 * 1 ST
       SFr. 776.00
  • Basic ESD 2 chair, made from Duotec ESD fabric, black, with contact backrest with tilting seat, backrest height: 530 mm
    470 - 610 mm
    Castors
    Five-legged base
    1 * 1 ST
    1 * 1 ST - SFr. 710.00
    SFr. 710.00
    1 * 1 ST
       SFr. 710.00
  • Basic ESD 2 chair, made from Skai® ESD synthetic leather, with contact backrest with tilting seat, backrest height: 530 mm
    470 - 610 mm
    Castors
    Five-legged base
    1 * 1 ST
    1 * 1 ST - SFr. 747.00
    SFr. 747.00
    1 * 1 ST
       SFr. 747.00
  • Basic ESD 2 chair, made from Duotec ESD fabric, blue, with contact backrest with tilting seat, backrest height: 530 mm
    470 - 610 mm
    Castors
    Five-legged base
    1 * 1 ST
    1 * 1 ST - SFr. 710.00
    SFr. 710.00
    1 * 1 ST
       SFr. 710.00
  • Basic ESD 2 chair, made from Duotec ESD fabric, black, with synchronous mechanism with weight regulation, backrest height: 530 mm
    470 - 610 mm
    Castors
    Five-legged base
    1 * 1 ST
    1 * 1 ST - SFr. 762.00
    SFr. 762.00
    1 * 1 ST
       SFr. 762.00
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..