HPLC columns for online SPE, HyperSep™ Hypercarb™
Supplier: Thermo Fisher Scientific
Certificates
About this item
When used with conventional HPLC systems and appropriate switching valves, these columns provide effective removal of contaminants, such as proteins, from samples and fast, effective clean-up and concentration of target compounds.
Frequently Bought Together