Orders LinkContactUsLinkComponent
Scintillation vials
Scintillation vials
Scintillation vials
Supplier:  DELTALAB
Certificates
Scintillation vials
2 Options Available Below
Product Details & Documents
Single use vials made of high density polyethylene to minimise solvent losses.
  • Screw cap ensures leakproof handling
  • Compatible with most liquid scintillation counters
Documents
Product Family Options
Product Information
  • Capacity
    Ø×H
    Cap included
    Cap type
    Availability
    Price
  • 4,00 ml
    13,71×53,15 mm
    No
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 435.00 SFr.
    435.00 SFr.
    1 * 2.000 ST
       435.00 SFr.
  • 20,00 ml
    26,5×58,5 mm
    Yes
    Screw cap
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 456.00 SFr.
    456.00 SFr.
    1 * 1.000 ST
       456.00 SFr.