0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Spray Caps
Spray pump for bottles
Spray pump for bottles
Spray pump for bottles
Catalog # MUEL14.289.14
Supplier:
MUELLER KREMPEL
Expand image
Spray pump for bottles
Catalog # MUEL14.289.14
Supplier:
MUELLER KREMPEL
Specifications
For:
vaporiser bottles 100 ml
Thread:
GL 20
Description:
Spray pump, white
Pk:
20
Specifications
About this item
Product Details
Product Details