ContactUsLinkComponent
Anion exchange HPLC columns, Dionex™, IonPac™ AS9-SC
Anion exchange HPLC columns, Dionex™, IonPac™ AS9-SC
Supplier:  DIONEX
Certificates
Anion exchange HPLC columns, Dionex™, IonPac™ AS9-SC
Supplier:  DIONEX
Specifications
  • Catalog No:
  • DION043186
  • DION043185
  • Max. pressure:
  • 4000 psi
  • 4000 psi
  • pH range:
  • 2 - 11
  • 2 - 11