Anion exchange HPLC columns, Dionex™, CarboPac™ MA1
: DIONEX
This column can be used for sensitive, rugged and reliable separations of reduced sugars without the need for derivatisation. This high capacity, strong anion exchange column efficiently separates reduced monosaccharides and disaccharides commonly found in food products and physiological samples.
- Catalog No:
- DION044066
- DION044067
- Max. pressure:
- 2000 psi
- 2000 psi
- pH range:
- 0 - 14
- 0 - 14
- Temperature limit:
- 4 - 50 °C
- 4 - 50 °C
Frequently Bought Together