Orders LinkContactUsLinkComponent
Human sera
Human sera
Supplier:   Biowest
Human sera
Supplier:  Biowest
Certificates
Human sera
5 Options Available Below
Product Details & Documents
Human serum 'off-the-clot' is processed from human blood that has been coagulation-induced. It is collected from volunteer donors.
Mainly sourced from France, Germany, Canada or USA, each batch is rigorously controlled and screened for Hepatitis B (HBS), Hepatitis C (HCV) and HIV Types 1 and 2 (HIV 1/2).
Important note: Products of human origin should be considered potentially infectious and handled accordingly.
Ordering information: Special treatments are available on request. Please contact your local Avantor Sales Office for more information.
Documents
Product Family Options
Product Information
  • Sterility
    Serum format
    Availability
    Price
  • S4190-100
    Human serum AB male, HIV tested
    Human serum AB male, HIV tested
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 389.00
    SFr. 389.00
    1 * 100 mL
       SFr. 389.00
  • S419G-100
    Human serum AB male, HIV tested, gamma irradiated
    Human serum AB male, HIV tested, gamma irradiated
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL
  • S419H-100
    Human serum AB male, HIV tested, heat inactivated
    Human serum AB male, HIV tested, heat inactivated
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 395.00
    SFr. 395.00
    1 * 100 mL
       SFr. 395.00
  • S4200-100
    Human serum, HIV tested
    Human serum, HIV tested
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 264.00
    SFr. 264.00
    1 * 100 mL
       SFr. 264.00
  • S420H-100
    Human serum, HIV tested, heat inactivated
    Human serum, HIV tested, heat inactivated
    Sterile filtered
    Frozen
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 270.00
    SFr. 270.00
    1 * 100 mL
       SFr. 270.00