Trypsin
About this item
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
16 Options Available Below
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Documents
Product Family Options
Product Information
- PkAvailabilityPrice
- L0909-100Trypsin 0,25% in PBS, without calcium, without magnesium, with phenol redTrypsin 0,25% in PBS, without calcium, without magnesium, with phenol red100 mLLog in for availability1 * 100 mLSFr. 16.60
- L0910-100Trypsin 0,25% in PBS, without calcium, without magnesium, without phenol redTrypsin 0,25% in PBS, without calcium, without magnesium, without phenol red100 mlLog in for availability1 * 100 mLSFr. 14.90
- X0915-100Trypsin 2,5% in PBS, without calcium, without magnesium, without phenol redTrypsin 2,5% in PBS, without calcium, without magnesium, without phenol red100 mlLog in for availability1 * 100 mLSFr. 26.50
- X0920-100Trypsin 2,5% in HBSS, without calcium, without magnesium, without phenol redTrypsin 2,5% in HBSS, without calcium, without magnesium, without phenol red100 mLLog in for availability1 * 100 mLSFr. 21.90
- L0930-100Trypsin-EDTA 1X in solution, without calcium, without magnesium, with phenol redTrypsin-EDTA 1X in solution, without calcium, without magnesium, with phenol red100 mlLog in for availability1 * 100 mLSFr. 12.30
- L0930-500Trypsin-EDTA 1X in solution, without calcium, without magnesium, with phenol redTrypsin-EDTA 1X in solution, without calcium, without magnesium, with phenol red500 mlLog in for availability1 * 500 mLSFr. 60.80
- L0940-100Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol redTrypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red100 mLLog in for availability1 * 100 mLSFr. 12.50
- L0940-500Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol redTrypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red500 mLLog in for availability1 * 500 mLSFr. 44.10
- P0940-1GRTrypsin-EDTA 1X, with sodium chloride, lyophilised
- P0940-100GRTrypsin-EDTA 1X, with sodium chloride, lyophilisedTrypsin-EDTA 1X, with sodium chloride, lyophilised100 gLog in for availability1 * 100 gSFr. 162.00
- X0930-100Trypsin-EDTA 10X
- L0931-100Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol redTrypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red100 mlLog in for availability1 * 100 mLSFr. 21.80
- L0931-500Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol redTrypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red500 mlLog in for availability1 * 500 mLSFr. 64.70
- L0932-100Trypsin 0,25% EDTA 0,02% in HBSS, without calcium, without magnesium, with phenol redTrypsin 0,25% EDTA 0,02% in HBSS, without calcium, without magnesium, with phenol red100 mLLog in for availability1 * 100 mLSFr. 26.50
- P5957-100GRTrypsin 1:250 powder (porcine)
- L0941-100Trypsin-EDTA (recombinant) 1X in PBS, without calcium, without magnesium, without phenol redTrypsin-EDTA (recombinant) 1X in PBS, without calcium, without magnesium, without phenol red100 mlLog in for availability1 * 100 mLSFr. 48.60