Order Entry
Switzerland
Orders LinkContactUsLinkComponent
ESD Saddle Chair

ESD Saddle Chair

Saddle chair, ESD, max. load 120 kg, 550 - 730 mm, Castors
Catalog # SOVE862010-00
Supplier:   TRESTON
ESD Saddle Chair
Catalog # SOVE862010-00
Supplier:  TRESTON
ESD Saddle Chair

Saddle chair, ESD, max. load 120 kg, 550 - 730 mm, Castors

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Chairs with freedom of movement with ergonomic sitting position.

  • Adjustment for seat height and seat tilt
  • Abrasion resistance >50000
  • Martindale with ESD textile
Specifications
  • Description:
    Saddle chair, ESD, max. load 120 kg
  • Height:
    550 - 730 mm
  • Foot style:
    Castors
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.