Order Entry
Switzerland
ContactUsLinkComponent
Cell culture dishes, poly-D-lysine, Corning® BioCoat™
Catalog # 734-0201
Anbieter:  Corning
   Low Manufacturing Impact
undefined
Cell culture dishes, poly-D-lysine, Corning® BioCoat™
Catalog # 734-0201
Anbieter:  Corning
   Low Manufacturing Impact

Technische Daten

  • Ø:
    150 mm
  • Growth area:
    151.9 cm²
  • Description:
    Culture dish, poly-D-lysine coated, with grid
  • Seq:
    07
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Pk:
    5

Technische Daten

Über diesen Artikel

TC-treated gridded culture dishes.

Corning® BioCoat™ products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.