Order Entry
Switzerland
ContactUsLinkComponent
Safety cutter, SECUPRO MARTEGO
Safety cutter, SECUPRO MARTEGO
Catalog # 223-0099
Supplier:  MARTOR ARGENTAX
undefined
Safety cutter, SECUPRO MARTEGO
Catalog # 223-0099
Supplier:  MARTOR ARGENTAX

Specifications

  • Total length:
    155 mm
  • Blade:
    26×18,50×0,4 mm
  • Pk:
    1

Specifications

About this item

Steel blade, aluminium handle.

  • For use by left and right-handed users
  • Spring-loaded blade retracts back automatically - even if the user continues to hold the squeeze lever
  • Safety lock prevents the blade from accidentally extending
  • Easy blade change - no additional tools required
  • With 2-edge/4-point blade for extra economy

This safety cutter with automatic blade retraction system is designed for all normal cutting jobs, especially box-slitting. The ergonomically designed squeeze lever - made out of aluminium and a soft plastic component - provides comfortable and non-slip handling.

Certifications: TÜV approved