Order Entry
Switzerland
ContactUsLinkComponent
Mini knee joint with cross section
Mini knee joint with cross section
Catalog # BINH1000170
Supplier:  3B Scientific
undefined
Mini knee joint with cross section
Catalog # BINH1000170
Supplier:  3B Scientific

Specifications

  • Description:
    Mini knee joint with cross section
  • Pk:
    1

Specifications

About this item

The mini knee joint has been reduced to a half of the natural size of a human knee but has kept all of the functionality of the larger knee joint models. In addition to the external anatomical structures of the knee, the medical or teaching professional now has the possibility to explain what is happening from 'within' the knee joint.

  • Mounted on the base

Dimensions: 10×14×24 cm

Weight: 0,35 kg