Order Entry
Switzerland
ContactUsLinkComponent
Accessories for TurbiDirect
Catalog # LOVI197655
Anbieter:  Lovibond
undefined
Accessories for TurbiDirect
Catalog # LOVI197655
Anbieter:  Lovibond

Technische Daten

  • Description:
    Turbidity vials with black lid (H = 55 mm; Ø = 24 mm)
  • Seq:
    910
  • Pk:
    12

Technische Daten

Über diesen Artikel