Order Entry
Switzerland
ContactUsLinkComponent
 
Super drum, omologated
Catalog # LAMA3209
Supplier:  LAMAPLAST
Super drum, omologated
Catalog # LAMA3209
Supplier:  LAMAPLAST

Specifications

  • Ext.Ø:
    750 mm
  • Capacity:
    335 L
  • Height:
    1070 mm
  • Pk:
    1

Specifications

About this item

These hermetic safety container are produced by rotational system in high density polyethylene with an high molecular weight. Suitable for the transport and storing of metal drums containing harmful, toxic substances.

  • Resistant to impact, chemical and atmospheric agents
  • Double wall lid with dutral seal and stainless steel iron ring for a perfect hermetic seal