Cervical spinal column
Référence BINH1000144
Fournisseur: 3B Scientific
Spécifications
- Weight:300 g
- Description:Cervical spinal column (19 cm)
- Pk:1
Spécifications
À propos de cet article
This real life replica of the anatomy of the cervical spinal column is a high quality model.
- Consisting of:
- Occipital plate
- Seven cervical vertebrae with intervertebral discs
- Cervical nerves
- Vertebral arteries
- Spinal cord
On a flexible stand.