Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Safety Containers

Safety Containers

11 l, 226×255 mm
Catalog # DELT240011
Supplier:   DELTALAB
Safety Containers
Catalog # DELT240011
Supplier:  DELTALAB
Safety Containers

11 l, 226×255 mm

Price
199.00 SFr.
Log in for availability
1 * 10 ST
Product Details & Documents

Safety containers, made of HDPE with lid, made of PP.

  • Suitable for solid waste type II and III, enable incineration
  • Container body consists hazard warning label

It consists two lids, an entire opening, taking advantage of the total diameter of the mouth of the container, allows to throw away bigger volume waste. A partial opening used for handling syringes, contact plates, and midsize waste

Certifications: In accordance with european standards ONU ADR, ISO 6706, BS 5404.

Documents

Specifications
  • Capacity:
    11 l
  • Ø ext.×H:
    226×255 mm