Order Entry
Switzerland
ContactUsLinkComponent
Examination light, Sovella® ProLite 28
Examination light, Sovella® ProLite 28
Catalog # SOVE861928-UK
Supplier:  TRESTON
undefined
Examination light, Sovella® ProLite 28
Catalog # SOVE861928-UK
Supplier:  TRESTON

Specifications

  • Description:
    Sovella®ProLite 28, 28 W, IP 20, 220-240 V, UK-plug
  • Pk:
    1

Specifications

About this item

This ergonomic lighting solution is environmentally friendly. Sovella® ProLite 28 is a standard light for workplaces where good general lighting is required but high accuracy is not necessary, for example packing stations.

  • Low energy consumption
  • All materials are recyclable
  • Prism diffuser shield which illuminates the work surface evenly and protects the fluorescent tube and technical components from dust

W×D×H: 1230×155×50 mm

Weight: 4,65 kg