Order Entry
Switzerland
ContactUsLinkComponent
Tongue depressor
Tongue depressor
Catalog # BAIL1105B100
Supplier:  BAILLY
Tongue depressor
Catalog # BAIL1105B100
Supplier:  BAILLY

Specifications

  • Width:
    18 mm
  • Description:
    Tongue depressor, for adults
  • Length:
    150 mm
  • Pk:
    100

Specifications

About this item

Wooden tongue depressor for adults.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904