ContactUsLinkComponent
Safety bulb pipette filler
Safety bulb pipette filler
Catalog # HACH1465100
Supplier:  Hach
undefined
Safety bulb pipette filler
Catalog # HACH1465100
Supplier:  Hach
Specifications
  • Colour:
    Red/white
  • Description:
    Safety bulb pipette filler, with cone adapter and autoclavable elastic wristband
  • Pk:
    1
Specifications
About this item
This cone adapter with rubber ball allows universal pipette filling.
  • Easy and safe
  • Autoclavable
  • Includes autoclavable elastic wristband