Orders LinkContactUsLinkComponent
Double bosshead
Double bosshead
Double bosshead
Certificates
Double bosshead
2 Options Available Below
Product Details & Documents
Double bosshead made from epoxy coated malleable iron.
Certifications: DIN 12895
Product Family Options
Product Information
  • For rod Ø
    Availability
    Price
  • JUCHVL04810
    Double bosshead with M8 brass thumb bolt
    Double bosshead with M8 brass thumb bolt
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 13.10
    SFr. 13.10
    1 * 1 ST
       SFr. 13.10
  • JUCHVL04810S
    Double bosshead with M8 stainless steel thumb bolt
    Double bosshead with M8 stainless steel thumb bolt
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 27.20
    SFr. 27.20
    1 * 1 ST
       SFr. 27.20