Orders LinkContactUsLinkComponent
Laboratory jacks
Laboratory jacks
Certificates
Laboratory jacks
Laboratory jacks
5 Options Available Below
Product Details & Documents
Aluminium alloy, with epoxy resin coating. These manually operated lifting stages provide high stablity.
  • Special epoxy resin coating ensures that the surfaces are extremely resistant to chemical attack and to other environmental factors
  • Strict quality controls, as required by DIN EN ISO 9001:2000, ensure a constant product quality
  • Made in Germany
Product Family Options
Product Information
  • Plate size
    Max. static load capacity
    Max. dynamic load capacity
    Working height
    Colour
    Availability
    Price
  • 100×100 mm
    60 kg
    4 kg
    60 - 200 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 70.80
    SFr. 70.80
    1 * 1 ST
       SFr. 70.80
  • 160×130 mm
    60 kg
    7 kg
    60 - 250 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 129.00
    SFr. 129.00
    1 * 1 ST
       SFr. 129.00
  • 200×230 mm
    60 kg
    7 kg
    70 - 260 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 167.00
    SFr. 167.00
    1 * 1 ST
       SFr. 167.00
  • 300×300 mm
    60 kg
    12 kg
    130 - 465 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 709.00
    SFr. 709.00
    1 * 1 ST
       SFr. 709.00
  • 400×170 mm
    60 kg
    15 kg
    170 - 615 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST