Order Entry
Switzerland
ContactUsLinkComponent
Syringe filter holders
Syringe filter holders
Nome del fornitore:  Merck
Certificati
undefined
Syringe filter holders
Nome del fornitore:  Merck

Specifiche

  • Catalog No:
  • MILFXX3002500
  • MILFXX3001200
  • MILFXX3002514
  • Filtration area:
  • 3,9 cm²
  • 0,8 cm²
  • 3,9 cm²
  • Inlet/outlet connections:
  • Inlet: Female Luer-Lok®
    Outlet: Male Luer-Slip™
  • Inlet: Female Luer-Lok®
    Outlet: Male Luer-Slip™
  • Inlet: 1/4" NPTF
    Outlet: Male Luer-Slip™
  • Max. operating pressure:
  • 6,9 bar
  • 6,9 bar
  • 6,9 bar

Specifiche

Frequently Bought Together