Cut resistant gloves, Edge® 16-500
Supplier: Ansell
Certificates
About this item
These interlock cotton liners feature a ³/₄ dipped natural rubber latex coating and a knitwrist cuff. They provide good cut resistance.
- Crinkle finish provides excellent wet and dry grip on slippery surfaces but also protects the hand from sharp edges
- Ergonomically designed to follow the natural shape of the hand: delivers a better fit
- Good cushioning, good cut resistance, good comfort
Ideal for handling glass, tiles, ceramics and slippery plastics, for waste collection, oyster-farming and working in green spaces.
Certifications: EN 388:2016 (X141B), EN 407 (X2XXXX); Cat. III.
Specifications
- Catalog No:
- ANSE16-500/8
- ANSE16-500/9
- ANSE16-500/7
- ANSE16-500/10
- Model:
- 16-500
- 16-500
- 16-500
- 16-500
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (X141B), EN 407 (X2XXXX)
- EN 388:2016 (X141B), EN 407 (X2XXXX)
- EN 388:2016 (X141B), EN 407 (X2XXXX)
- EN 388:2016 (X141B), EN 407 (X2XXXX)