Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Human skull model

Human skull model

Human skull model in five parts, 1,1 kg, 200×135×155 mm
Catalog # BINH1000049
Marketsource
Supplier:   3B Scientific
Human skull model
Catalog # BINH1000049
Supplier:  3B Scientific
Marketsource
Human skull model
Human skull model

Human skull model in five parts, 1,1 kg, 200×135×155 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

The three-part medical quality human skull replica is designed for basic anatomical studies of the skull. It includes an anatomically correct five-part brain.

  • High quality original skull cast
  • Handmade from hard plastic skull
  • Highly accurate representation of the fissures, foramina, processes, sutures
  • Can be disassembled into skull cap, base of skull, and mandible
  • Mandible of skull is mounted on a spring to easily demonstrate natural movement

This medical grade skull model also makes a great present for medical professionals and students of medicine and allied health professions.

Specifications
  • Weight:
    1,1 kg
  • Description:
    Human skull model in five parts
  • W×D×H:
    200×135×155 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.