ContactUsLinkComponent
Accessories for orbital shakers
Accessories for orbital shakers
  444-0521
 :  IKA
undefined
Accessories for orbital shakers
  444-0521
 :  IKA
  • Description:
    Universal adhesive mat, STICKMAX, (W×D): 200×200 mm, max. speed 300 rpm
  • Pk:
    1
  • For:
    KS 4000 i control, KS 4000 ic control, KS 3000 i control, KS 3000 ic control, KS 130 basic, KS 130 control, KS 130 control NOL, KS 260 basic, KS 260 control, KS 260 control NOL, HS 501 digital