Bouffant dispenser with lid
Catalog # SCURBD-600
Supplier: S-CURVE
Specifications
- Dispenser type:
- PPE type:
- Colour:Clear
- Description:Dispenser for bouffant caps, with hinged lid and front large round opening, 1 compartment, acrylic
- W×D×H:304,8×203,2×304,8 mm
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Bouffant dispenser, made of clear acrylic, for cleanroom bouffants.
- Single compartment
- Lid with full length piano hinge
- Large round opening for easy access
- Flame treated edges for a smooth finish
- Keyholes for wall mounting