Lab Markers, several colours
Supplier: PRECISION DYNAMICS
Certificates
About this item
Versatile, permanent markings write on plastic, metal, and porcelain labware.
- Several colours
- Unique ink has the ability to write on labware even when the surface is damp
Frequently Bought Together