Heat and cut resistant gloves
Anbieter: JUTEC
Zertifikate
Über diesen Artikel
These five-finger gloves in Kevlar® coarse knit with cotton insulation protect against contact heats of up to 250 °C.
- Perfect for work involving slight heat influence
- The gloves are single-insulated and offer excellent handling
- Washable
- With knitted cuff
The gloves can be used in any environment in which direct exposure to flames is ruled out from the outset.
Certifications: EN 388 (244X), EN 407 (X1XXXX); Cat. II.
Technische Daten
- Catalog No:
- JUTEH0150035
- 112-1962
- Disposable / reusable:
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Liner material:
- Cotton
- Cotton
- Cuff style:
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (234X), EN 407 (X1XXXX)
- EN 388 (234X), EN 407 (X1XXXX)
Technische Daten
Frequently Bought Together