ContactUsLinkComponent
Protective barrier cream, Stokoderm® Universal PURE
Protective barrier cream, Stokoderm® Universal PURE
Catalog # 115-2250
Supplier:  Deb-STOKO
undefined
Protective barrier cream, Stokoderm® Universal PURE
Catalog # 115-2250
Supplier:  Deb-STOKO
Specifications
  • Description:
    Protective barrier cream, Stokoderm® Universal PURE, perfume-free, 1000 ml bottle
  • Pk:
    9
Specifications
About this item
Triple action cream protects against water and oil based substances, plus helps improve grip and comfort under gloves. Perfume-free and dye-free.
  • Unique triple action formulation
  • Contains Zinc Salicylate astringent agent and a blend of skin caring ingredients
  • Proven general purpose protection and glove compatibility
  • Formulated with a low pH value (4.7-5.2)
  • Hypoallergenic
  • Silicone-free
Suitable for most workplaces to protect against oil-based and water-based substances (such as oil, grease, lubricants, and detergents) and where gloves are worn. Apply 1ml to clean and dry skin before starting each work period and after washing hands. Rub on all areas of the hands, including between the fingers and around the fingernails.
Certifications: This product meets the European Centre for Allergy Research Foundation (ECARF) criteria for skin tolerability.