Orders LinkContactUsLinkComponent
Pipette tips, BagTips®
Pipette tips, BagTips®
Pipette tips, BagTips®
Supplier:  INTERSCIENCE
Certificates
Pipette tips, BagTips®
Pipette tips, BagTips®
2 Options Available Below
Product Details & Documents
These sterile pipette tips are designed for use with the BagPipet® pipette. They are available in two sizes: Regular (19 cm) and Jumbo (24 cm).
Product Family Options
Product Information
  • Tip style
    Sterile
    Length
    For use with
    Packed
    Availability
    Price
  • Straw type, regular
    +
    190 mm
    BagPipet® pipette
    1000/box
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 55.80
    SFr. 55.80
    1 * 1.000 ST
       SFr. 55.80
  • Straw type, jumbo
    +
    240 mm
    BagPipet® pipette
    1000/box
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 60.90
    SFr. 60.90
    1 * 1.000 ST
       SFr. 60.90