ContactUsLinkComponent
Single channel pipettes, mechanical, BagPipet®
Single channel pipettes, mechanical, BagPipet®
Catalog # 432-0012
Supplier:  INTERSCIENCE
undefined
Single channel pipettes, mechanical, BagPipet®
Catalog # 432-0012
Supplier:  INTERSCIENCE
Specifications
  • Volume:
    0,1/0,9/1 ml
  • Description:
    Autoclavable pipette, for use with BagTips®
  • Pk:
    1
Specifications
About this item
BagPipet® is an autoclavable pipette with three selectable volumes (0,1 ml / 0,9 ml / 1 ml). It is used with BagTips®, a single-use sterile and rigid straw.
  • Economical
  • No risk of cross-contamination
  • Light and ergonomic
  • Autoclavable and calibrated according to the GLP