Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Replica-plating device, SP Bel-Art

Replica-plating device, SP Bel-Art
Certificates
Replica-plating device, SP Bel-Art
Replica-plating device, SP Bel-Art

2 Options Available Below

Product Details & Documents

For colony replication, the Petri dish with the bacterial colonies in it is overturned, pressed into the fabric surface of the colony replicating platform and then removed. A replica of the original colonies (now sitting on the replicating platform) is then obtained by inverting a sterile Petri plate with appropriate nutrient agar in it and pressing it into the velveteen surface of the replicating platform.

  • Replicating device is easily disinfected after every use with ethyl alcohol or chlorine bleach
  • Can be used with 90 to 100 mm Petri dishes
  • Includes one bag of 12 sterile 100% cotton velveteen squares (152×152 mm)
  • Accurate, economical colony replication

Dimensions: Aluminium locking ring Ø 102 mm; PVC cylinder height 69 mm.

 : Replacement velveteen squares are available packed in a sealed bag with three inner locking bags each containing 12 squares (36 pieces in total).

Product Family Options

Product Information

  • Availability
    Price
  • 601-3401
    Replica-plating device, consisting of aluminium locking ring (Ø 102 mm) and PVC cylinder (69 mm)
    Replica-plating device, consisting of aluminium locking ring (Ø 102 mm) and PVC cylinder (69 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 345.00 SFr.
    345.00 SFr.
    1 * 1 ST
       345.00 SFr.

  • 601-3402
    Velveteen squares, twill-lined, white, 100% cotton, porous, sterile, 152×152 mm
    Velveteen squares, twill-lined, white, 100% cotton, porous, sterile, 152×152 mm
    Log in for availability
    Log in for availability
    1 * 36 ST
    1 * 36 ST - 406.00 SFr.
    406.00 SFr.
    1 * 36 ST
       406.00 SFr.