ContactUsLinkComponent
Glass cutter
Glass cutter
  272-9015
undefined
Glass cutter
  272-9015
  • Description:
    Glass cutter with 6 small steel wheels
  • Pk:
    1
Wooden handle with special steel wheels.
Glass cutter with six special steel wheels for cutting glass plates, TL-plates, microslides, etc.