Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bossheads

Bossheads

2 - 16 mm, Aluminium alloy
Catalog # 241-3001
Supplier:   BOCHEM
Bossheads
Catalog # 241-3001
Supplier:  BOCHEM
Bossheads
Bossheads

2 - 16 mm, Aluminium alloy

Price
SFr. 55.60
Log in for availability
1 * 1 ST
Product Details & Documents

Bossheads made of laboral (aluminium alloy).

  • For rods up to 16 mm
  • Angle: 90°
Specifications
  • Spanning width:
    2 - 16 mm
  • Material:
    Aluminium alloy
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.