Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Burette stand, DUPLEX

Burette stand, DUPLEX

Burette stand, green cast iron, incl. rod (600×10 mm) and two plates (Ø 105 mm), 2 kg, Two burettes
Catalog # BOCH5620
Supplier:   BOCHEM
Burette stand, DUPLEX
Catalog # BOCH5620
Supplier:  BOCHEM
Burette stand, DUPLEX
Burette stand, DUPLEX

Burette stand, green cast iron, incl. rod (600×10 mm) and two plates (Ø 105 mm), 2 kg, Two burettes

Price
SFr. 127.00
Log in for availability
1 * 1 KIT
Product Details & Documents

This burette stand is made of green varnished cast iron.

  • Incl. stainless steel rod (600×10 mm)
  • Incl. two plates with diameter 105 mm (black and white)
  • For use with burette holder (article no. 5600)
Specifications
  • For:
    Two burettes
  • Description:
    Burette stand, green cast iron, incl. rod (600×10 mm) and two plates (Ø 105 mm)
  • Weight:
    2 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.