Reagent reservoirs, Axygen®
: Corning
PS, white. These disposable reservoirs are designed to accommodate all multi channel pipettors.
- Certified RNase-/DNase-free
- Non pyrogenic
- Four corners and sharp V-bottom shape ensures maximum sample retrieval
- Available sterile or non sterile
- 25 ml reservoir also available with two channels (divided)
- Catalog No:
- AXYGRES-V-50-S
- AXYGRES-2CV-25-S
- AXYGRES-2CV-25-SI
- AXYGRES-V-25-SI
- AXYGRES-V-50-SI
- AXYGRES-2CV-25
- AXYGRES-V-25
- AXYGRES-V-25-S
- AXYGRES-V-100-S
- AXYGRES-V-100-SI
- AXYGRES-V-50
- AXYGRES-V-100
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Frequently Bought Together