ContactUsLinkComponent
Manual compression sealer for AxyMat™ sealing mats, Axygen® EASY PRESS
  732-0907
 :  Corning
   Low Manufacturing Impact
undefined
Manual compression sealer for AxyMat™ sealing mats, Axygen® EASY PRESS
  732-0907
 :  Corning
   Low Manufacturing Impact
  • Description:
    Axygen® EASY PRESS, manual compression sealer for AxyMat™ sealing mats
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Pk:
    1
Axygen® EASY PRESS is a manual device which provides simple and effective application of Axymat™ and/or Impermamat™ sealing mats to 24, 48, 96 or 384 well microplates, deep well plates, microtitre tube systems and PCR plates. The EASY PRESS can be quickly adjusted to handle any size plate of varying heights (384 PCR to 2 ml deep well blocks) using the large thumbwheel.