Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Melatonin

Melatonin

Melatonin
Supplier:  Cayman Chemical
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Formula:C₁₃H₁₆N₂O₂
  • MDL Number:MFCD00005655
  • Molecular Weight:232.28 g/mol
  • EINECS:200-797-7

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 68.40
    SFr. 68.40
    1 * 5 g
       SFr. 68.40

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - SFr. 130.00
    SFr. 130.00
    1 * 10 g
       SFr. 130.00