J.T.Baker®, Tantalum Standard Solution, 10000 mg/L Ta in Water with Hydrofluoric Acid (max. 1%) (from TaCl5) for ICP
Catalog # 5748.0100
Supplier: Avantor
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:H₂O/tr HF
- Pack type:Glass bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Tantalum
- Symbol:Ta