2-Cyclopentylethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:766-00-7
:
Formula:C₇H₁₄O
MDL Number:MFCD00019301
:114.19 g/mol
Boiling Pt:182°C
Flash Pt:70°C
EINECS:212-156-9
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together