

4-Phenyl-3-butyn-2-ol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5876-76-6
:
Formula:C₁₀H₁₀
MDL Number:MFCD00046651
:146.19 g/mol
Density:1.005
Flash Pt:>110 °C (230 °F)
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together